Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IgG, Sheep anti-Human, Cy3, Polyclonal,

Catalog No. AC112CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Cyanine3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AC112CMI 100 μL Cyanine3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112CMI Supplier MilliporeSigma™ Supplier No. AC112C
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Immunoglobulin G (IgG) is part of the family of glycoprotein immunoglobulins, which stage immune responses against pathogenic organisms. IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. IgG contributes to the immune response by mobilizing innate immune effector cells and inducing and pro-inflammatory responses. Such proinflammatory responses are mediated by Fc gamma receptors which interact with sialylated Fc fragments on IgG molecules. The coupling of Cy3 to Anti-Human IgG antibody allows for the visualization of proteins by fluorescent microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Cyanine3
Formulation Purified sheep polyclonal antibody Cy3 conjugate in PBS with 1.5% BSA and 0.09% Sodium Azide.
Host Species Sheep
Immunogen Purified human IgG.
Purification Method Ammonium Sulfate
Quantity 100 μL
Regulatory Status RUO
Research Discipline Secondary & Control Antibodies
Primary or Secondary Primary
Test Specificity Based on ELISA analysis using the unconjugated antibody (Mfr. No. AC112), this antibody exhibits strongest immunoreactivity towards whole human IgG molecule (IgG>IgM>IgG Fc>IgG Fab>IgA>IgM Fc) with minimal or no cross-reactivity toward IgG from chicken, guinea pig, hamster, mouse, rabbit, and rat species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.