Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, Sheep, Unconjugated, Polyclonal, Isotype Control, Novus Biologicals™
SDP

Catalog No. NBP19705510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19705510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19705510 Supplier Novus Biologicals Supplier No. NBP19705510MG
Add to Cart
Add to Cart

Sheep Antibody

IgG specifically detects IgG in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page.

Specifications

Antigen IgG
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen, SDS-Page 1:100 - 1:2000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Host Species Sheep
Purification Method Ion exchange chromatography
Quantity 10 mg
Primary or Secondary Isotype Control (Primary)
Test Specificity Sheep IgG whole molecule was assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Sheep IgG and anti-Sheep Serum.
Reconstitution Reconstitute with 1.0 mL deionized water.
Target Species Sheep
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.