Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IgG, Species absorbed, Sheep anti-Mouse, Biotin, Polyclonal,

Catalog No. AC111BMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AC111BMI 200 μL Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111BMI Supplier MilliporeSigma™ Supplier No. AC111B
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Immunoglobulin G (IgG) is part of the family of glycoprotein immunoglobulins which stage immune responses against pathogenic organisms. IgG contributes to the immune response by mobilizing innate immune effector cells and inducing and pro-inflammatory responses. Such proinflammatory responses are mediated by Fcg receptors which interact with sialylated Fc fragments on IgG molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG, Species absorbed
Applications Immunohistochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Biotin
Formulation Purified sheep polyclonal antibody in PBS with 15 mg/mL BSA and 0.1 % Sodium Azide.
Host Species Sheep
Immunogen Purified mouse IgG.
Purification Method Fractionated by octanoic acid and ammonium sulfate cut, and then depleted with rat IgG column and mouse IgA column prior to biotin conjugation.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Secondary & Control Antibodies
Primary or Secondary Primary
Test Specificity Based on ELISA analysis using the unconjugated antibody (Mfr. No. AC111), this antibody reacts with whole molecule mouse IgG (IgG2 IgG2a IgG3 IgG1) with minimal or no cross-reactivity toward mouse IgA or IgM.
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.