Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1 Antibody (SR1920), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322442100UL
Description
IgG1 Monoclonal antibody specifically detects IgG1 in Human samples. It is validated for Western BlotSpecifications
IgG1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Human | |
Purified |
Western Blot | |
SR1920 | |
Western Blot 1:500-1:2000 | |
IgG1, Igh-4, RGD1359539, VH7183 | |
A synthesized peptide derived from human IgG1 (Uniprot #: P01857) | |
100 μg | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction