Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Kappa Light Chain Rat, Allophycocyanin, Clone: KLH/G1-2-2, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB001935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001935 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001935 Supplier Novus Biologicals Supplier No. NBP151817APC500UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG1 Kappa Light Chain Monoclonal specifically detects IgG1 Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, ELISA, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG1 Kappa Light Chain
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone KLH/G1-2-2
Conjugate APC
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Formulation PBS.
Host Species Rat
Immunogen KLH
Quantity 500 μL
Test Specificity Immunoglobulin reacts with Keyhole limpet hemocyanin (KLH).
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.