Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Kpa ItCl Armenian Hamster, R718, Clone: A19-3, Isotype Control, BD Horizon™

Catalog No. BDB566951 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566951 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566951 Supplier BD Biosciences Supplier No. 566951
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The A19-3 monoclonal antibody specifically binds to the hapten, trinitrophenol (TNP). TNP is not expressed on human, mouse, or rat cells. The immunoglobulin from clone A19-3 was selected as an isotype control following screening for low background staining on a variety of mouse and human tissues.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1 Kpa ItCl
Applications Flow Cytometry
Classification Monoclonal
Clone A19-3
Conjugate R718
Host Species Armenian Hamster
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.