Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Mouse, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB170775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170775 500 μg
NB170774 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170775 Supplier Novus Biologicals Supplier No. NBP318109500UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Isotype Control

IgG1 Monoclonal antibody specifically detects IgG1 in samples. It is validated for Control

Specifications

Antigen IgG1
Classification Monoclonal
Clone B-D38
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Mouse
Immunogen Chemical molecule. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Immunology
Content And Storage Store at 4°C. Do not freeze.
Product Type Isotype Controls
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.