Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 PAb, anti-Mouse, Rabbit Affin. Purif. Liq., FITC, MP Biomedicals

Catalog No. ICN50260 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN50260 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN50260 Supplier MP Biomedicals, Inc Supplier No. 50260

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC
Quantity 2 mL
Primary or Secondary Primary
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.