Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2 Antibody (SR1938), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | IgG2 |
---|---|
Clone | SR1938 |
Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
IgG2 | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker) | |
A synthesized peptide derived from human IgG2 (Uniprot #: P01859) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1938 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Immunology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3501 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title