Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2 Human, Alexa Fluor 647, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB183515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183515 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183515 Supplier Novus Biologicals Supplier No. NBP306870AF647
This item is not returnable. View return policy

Human Monoclonal Isotype Control

IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Control

Specifications

Antigen IgG2
Classification Monoclonal
Clone MAB1641
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker)
Host Species Human
Immunogen Human myeloma serum
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Gene ID (Entrez) 3501
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Isotype Controls
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.