Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2 Human, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB183507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183507 0.1 mg
NB183508 0.5 mg
NB183509 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB183507 Supplier Novus Biologicals Supplier No. NBP3068700.1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Monoclonal Isotype Control

IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Control

Specifications

Antigen IgG2
Classification Monoclonal
Clone MAB1641
Conjugate Unconjugated
Formulation Lyophilized from a 0.2 um filtered solution in PBS. 0.5mg and 0.1 mg sizes are provided in liquid form, PBS
Gene Alias Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker)
Host Species Human
Immunogen Human myeloma serum
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Gene ID (Entrez) 3501
Reconstitution Reconstitute with sterile 1 mL PBS
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Isotype Controls
Form Lyophilized
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.