Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2 Mouse anti-Human, BV650, Clone: HP6002 (also known as AB4-DH4), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756050
Description
IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Flow cytometry
Specifications
IgG2 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
3501 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
HP6002 (also known as AB4-DH4) | |
Brilliant Violet 650 | |
IGHG2; Immunoglobulin heavy constant gamma 2 | |
Purified Human IgG2 Myeloma Protein | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction