Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2 Mouse anti-Human, PE, Clone: HP6002 (also known as AB4-DH4), BD Pharmingen™

Catalog No. BDB568959 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568959 100 Tests
BDB568960 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB568959 Supplier BD Biosciences Supplier No. 568959
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Flow cytometry (Routinely Tested)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2
Applications Flow Cytometry
Classification Monoclonal
Clone HP6002 (also known as AB4-DH4)
Conjugate PE
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Gene Alias IGHG2; Immunoglobulin heavy constant gamma 2
Host Species Mouse
Immunogen Purified Human IgG2 Myeloma Protein
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3501
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.