Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2a/IgG2b Mouse, R718, Clone: R2-40, Isotype Control, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752047
Description
The R2-40 monoclonal antibody specifically recognizes a common epitope, probably located in the CH1 domain, shared by mouse IgG2a, and IgG2b, of Igh-Ca and Igh-Cb haplotypes. It does not react with other Ig isotypes.

Specifications
IgG2a/IgG2b | |
Monoclonal | |
R718 | |
50 μg | |
Isotype Control (Primary) | |
Liquid |
Flow Cytometry | |
R2-40 | |
Mouse | |
RUO | |
Refrigerate Upon Arrival |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction