Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2a kappa Isotype Control (eBR2a), Biotin, eBioscience™, Invitrogen™

Rat Isotype Control

$83.00 - $377.00

Specifications

Antigen Rat IgG2a kappa
Clone eBR2a
Concentration 0.5 mg/mL
Applications Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Conjugate Biotin
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-9598
Encompass_Preferred
View Documents
Invitrogen
13432181
50 μg
Each for $83.00
Only null left
Edge
Add to Cart
 
50-112-9638
Encompass_Preferred
View Documents
Invitrogen
13432182
100 μg
Each for $153.00
Only null left
Edge
Add to Cart
 
50-118-57
Encompass_Preferred
View Documents
Invitrogen
13432185
500 μg
Each for $377.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in immunocytochemistry, immunohistochemistry, flow cytometric analysis, and ELISA. Applications Tested: This rat IgG2a isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and should be used at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.
Specifications

Specifications

Rat IgG2a kappa
0.5 mg/mL
Biotin
Rat
Not Applicable
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
eBR2a
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
IgG2a κ
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.