Learn More
Rat IgG2a kappa Isotype Control (eBR2a), FITC, eBioscience™, Invitrogen™
Rat Isotype Control
$73.25 - $283.00
Specifications
Antigen | Rat IgG2a kappa |
---|---|
Clone | eBR2a |
Applications | Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry |
Conjugate | FITC |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-186-07
![]() |
Invitrogen
11432180 |
25 μg |
Each for $73.25
|
|
|||||
50-112-2122
![]() |
Invitrogen
11432181 |
50 μg |
Each for $114.00
|
![]() |
|||||
50-112-9602
![]() |
Invitrogen
11432182 |
100 μg |
Each for $184.00
|
![]() |
|||||
50-186-10
![]() |
Invitrogen
11432185 |
500 μg |
Each for $283.00
|
|
|||||
50-186-05
![]() |
Invitrogen
11432141 |
25 Tests |
Each for $111.00
|
|
|||||
50-186-06
![]() |
Invitrogen
11432142 |
100 Tests |
Each for $195.00
|
![]() |
|||||
Description
Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: The FITC conjugated rat IgG2a isotype control is offered in 2 formats: - μg size: This rat IgG2a isotype control has been tested by flow cytometric analysis of mouse splenocytes. This can be used at the same concentration as the experimental antibody. - test size: has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells and can be used at 5 μL for the catalog number ending in -41 or -42 and 20μL/test for the product ending in -73 per test. Each test size contains 0.5 μg. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.Specifications
Rat IgG2a kappa | |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
Liquid | |
RUO | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBR2a | |
FITC | |
Rat | |
Not Applicable | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.