Learn More
BD Horizon™ IgG2a κ Mouse, RY586, Clone: G155-178, Isotype Control, BD Horizon™
Shop All BD Biosciences ProductsDescription
The G155-178 clone has an unknown specificity. Trinitrophenol (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to immunoglobulin Fc receptors. The immunoglobulin secreted by the G155-178 hybridoma was selected as a mouse IgG2a, κ isotype control following screening for low background binding on a variety of mouse and human tissues.
Specifications
Specifications
| Antigen | IgG2a κ |
| Applications | Flow Cytometry |
| Clone | G155-178 |
| Conjugate | RY586 |
| Host Species | Mouse |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Isotype Control (Primary) |
| Isotype | IgG2a κ |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.