Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2a Mouse, Unconjugated, Clone: M2AK, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB001273 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001273 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001273 Supplier Novus Biologicals Supplier No. NBP1969810.5MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG2a Kappa Monoclonal specifically detects IgG2a Kappa in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG2a
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoelectrophoresis
Classification Monoclonal
Clone M2AK
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 1:1000-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation, SDS-Page, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Formulation 0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2
Host Species Mouse
Quantity 0.5 mg
Test Specificity In an Ouchterlony double diffusion assay the material is non-reactive with antisera to mouse IgG1, IgG2b, IgG3, IgM, and IgA. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse IgG and anti-Mouse serum.
Target Species Mouse
Content And Storage Store at 4°C short term. For extended storage, add an equal volume of glycerol aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.