Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2b Kpa ItCl Rat, R718, Clone: R35-38, Isotype Control, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566943
Description
The R35-38 hybridoma produces monoclonal rat IgG2b, κ. The target molecule specificity of this clone is unknown. R35-38 has little or no reactivity with mammalian cells and has been found useful as an immunoglobulin isotype control.

Specifications
IgG2b Kpa ItCl | |
Monoclonal | |
R718 | |
50 μg | |
Isotype Control (Primary) | |
Liquid |
Flow Cytometry | |
R35-38 | |
Rat | |
RUO | |
Refrigerate Upon Arrival |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction