Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2b Mouse, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB170773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170773 500 μg
NB170772 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170773 Supplier Novus Biologicals Supplier No. NBP318108500UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Isotype Control

IgG2b Monoclonal antibody specifically detects IgG2b in samples. It is validated for Control

Specifications

Antigen IgG2b
Classification Monoclonal
Clone B-F33
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS. Sterile-filtered through 0.22 um.
Host Species Mouse
Immunogen Chemical molecule. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells
Purification Method Affinity purified
Quantity 500 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Content And Storage Store at 4°C. Do not freeze.
Product Type Isotype Controls
Form Purified
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.