Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2c Kappa Rat, Janelia Fluor 549, Isotype Control, Novus Biologicals™
SDP

Catalog No. NB170420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170420 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB170420 Supplier Novus Biologicals Supplier No. NBP231387JF549
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG2c Kappa Monoclonal antibody specifically detects IgG2c Kappa in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, ELISA

Specifications

Antigen IgG2c Kappa
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, ELISA
Classification Monoclonal
Clone RG2cK
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Host Species Rat
Purification Method Protein G purified
Quantity 0.5 mL
Regulatory Status RUO
Target Species Rat
Content And Storage Store at 4°C in the dark.
Product Type Isotype Controls
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.