Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG3 Antibody (008), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290253AF700
Description
IgG3 Monoclonal specifically detects IgG3 in Human samples. It is validated for ELISA.Specifications
IgG3 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
8 | |
ELISA | |
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IgG3 (P01860; Glu99-Lys377). | |
0.1 mL | |
Immunology | |
3502 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction