Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG3 Antibody (SR1690), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321815100UL
Description
IgG3 Monoclonal antibody specifically detects IgG3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
IgG3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR1690 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
A synthesized peptide derived from human IgG3 (Uniprot #: P01860) | |
100 μg | |
Immunology | |
3502 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction