Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG3 Mouse anti-Human, Clone: MTG34, Novus Biologicals™

Mouse Monoclonal Antibody
$335.78 - $1148.45
Specifications
| Antigen | IgG3 |
|---|---|
| Clone | MTG34 |
| Concentration | 0.5 mg/ml |
| Applications | ELISA |
| Classification | Monoclonal |
Description
IgG3 Monoclonal antibody specifically detects IgG3 in Human samples. It is validated for ELISASpecifications
| IgG3 | |
| 0.5 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
| Human IgG3 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| MTG34 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| 0.2 um filtered PBS | |
| 3502 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title