Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG3 Mouse anti-Human, PE, Clone: HP6050 (also known as CC4-DC10), BD Pharmingen™
Mouse Monoclonal Antibody
$158.00 - $355.50
Specifications
| Antigen | IgG3 |
|---|---|
| Clone | HP6050 (also known as CC4-DC10) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
Description
IgG3 Monoclonal antibody specifically detects IgG3 in Human samples. It is validated for Flow CytometrySpecifications
| IgG3 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| IGHG3; Ig gamma-3 chain C region; Immunoglobulin heavy constant gamma 3 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
| HP6050 (also known as CC4-DC10) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human IgG3 myeloma protein | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title