Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG3, κ, Mouse, BUV737, Clone: J606, Isotype Control, BD Horizon™

Catalog No. BDB565360 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565360 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565360 Supplier BD Biosciences Supplier No. 565360

Mouse Monoclonal Isotype Control

IgG3, κ Monoclonal antibody specifically detects IgG3, κ in samples. It is validated for Flow cytometry, Isotype control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG3, κ
Applications Flow Cytometry
Classification Monoclonal
Clone J606
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 737
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Anti-Fructosan
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Form Purified
Isotype IgG3 κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.