Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG4 Antibody (023), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290223APC
Description
IgG4 Monoclonal antibody specifically detects IgG4 in Human samples. It is validated for ELISASpecifications
IgG4 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Human | |
Purified |
ELISA | |
023 | |
ELISA | |
Immunoglobulin G, Immunoglobulin G4, ImmunoglobulinG | |
Rabbit immunized with purified, recombinant Human IgG4 (Uniprot#: P01861, Glu99-Lys327,108Ser/Pro) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction