Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG4 Antibody (023), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290223V
Description
IgG4 Monoclonal specifically detects IgG4 in Human samples. It is validated for ELISA.Specifications
IgG4 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Human | |
Purified |
ELISA | |
23 | |
ELISA | |
Immunoglobulin G, Immunoglobulin G4, ImmunoglobulinG | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IgG4 (P01861; Glu99-Lys327,108Ser/Pro). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction