Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgGF(ab)2 Pig, Biotin, Isotype Control, Novus Biologicals™
SDP

Catalog No. NBP19681005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19681005 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP19681005 Supplier Novus Biologicals Supplier No. NBP1968100.5MG

Pig Antibody

IgG F(ab)2 specifically detects IgG F(ab)2 in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen IgGF(ab)2
Applications ELISA, Immunohistochemistry, Western Blot
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 0.02M Potassium Phosphate, 0.5M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Host Species Pig
Purification Method Ion exchange chromatography
Quantity 0.5 mg
Primary or Secondary Isotype Control (Primary)
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Porcine IgG, anti-Porcine IgG F(ab)2 and anti-Porcine Serum. No reaction was observed against anti-Porcine IgG F(c) or anti-Pepsin.
Reconstitution Reconstitute with 1.0mL deionized water.
Target Species Pig
Content And Storage Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.