Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgGF(ab) Chicken, Isotype Control, Novus Biologicals™
SDP

Catalog No. NBP1967732 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1967732 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP1967732 Supplier Novus Biologicals Supplier No. NBP1967732MG
Add to Cart
Add to Cart

Chicken Antibody

IgG F(ab) specifically detects IgG F(ab) in Chicken samples. It is validated for Western Blot, ELISA, SDS-Page.

Specifications

Antigen IgGF(ab)
Applications ELISA, Western Blot
Concentration 2mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000
Formulation 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Host Species Chicken
Purification Method Ion exchange chromatography
Quantity 2 mg
Primary or Secondary Isotype Control (Primary)
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Chicken IgG, anti-Chicken IgG F(ab)2 and anti-Chicken Serum. No reaction was observed against anti-Chicken IgG F(c) or anti- Papain.
Target Species Chicken
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.