Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGHA1 Mouse anti-Human, Monkey, Mouse, Clone:7D12C5, Abnova™

Catalog No. 89334466 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-466 Supplier Abnova Corporation Supplier No. MAB17823
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human IGHA1.

Specifications

Antigen IGHA1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7D12C5
Conjugate Unconjugated
Description immunoglobulin heavy constant alpha 1
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry (1:200-1:1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene IGHA1
Gene Alias FLJ14473, FLJ35065, FLJ35500, FLJ36402, FLJ39698, FLJ40001, FLJ41548, FLJ41552, FLJ41789, FLJ43248, FLJ43594, FLJ44293, FLJ46028, FLJ46621, FLJ46724, FLJ46811, FLJ46824, IgA1, MGC102857
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 207-353 of human IGHA1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 3493
Target Species Human, Monkey, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.