Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGHG1 Rabbit anti-Human, Clone-IG1707R, Novus Biologicals™

Rabbit Monoclonal Antibody
$646.00
Specifications
Antigen | IGHG1 |
---|---|
Clone | IG1707R |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254449100
![]() |
Novus Biologicals
NBP254449100UG |
100 μg |
Each for $646.00
|
|
|||||
NB254449200
![]() |
Novus Biologicals
NBP254449200UG |
200 μg | N/A | N/A | N/A | ||||
Description
IgG Heavy Chain Monoclonal specifically detects IgG Heavy Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGHG1 | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
RUO | |
Human | |
immunoglobulin heavy constant gamma 1 (G1m marker) | |
IGHG1 | |
IgG κ | |
Protein A purified | |
75 kDa |
IG1707R | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Rabbit | |
Immunology | |
PBS with No Preservative | |
3500 | |
Purified human Ig Gamma Chain | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title