Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGHG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571418 Supplier Invitrogen™ Supplier No. PA571418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGHG3 Polyclonal Antibody for Western Blot

IGHG3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGHG3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IGHG3
Gene Accession No. P01860
Gene Alias IgG3; IGHG3; immunoglobulin heavy constant gamma 3 (G3m marker)
Gene Symbols IGHG3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human IGHG3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3502
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.