Learn More
IGHM Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 110161AP150UL
Description
This antibody detects the heavy chain of human IgM (∽75 kDa).
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.Specifications
| IGHM | |
| Polyclonal | |
| Unconjugated | |
| IGHM | |
| IGHM, MU, VH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3507 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P01871 | |
| IGHM | |
| IGHM Fusion Protein Ag1459 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.