Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGHMBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103794 Supplier Thermo Scientific Supplier No. PA5103794
Add to Cart
Add to Cart

IGHMBP2 Polyclonal Antibody

IGHMBP2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen IGHMBP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IGHMBP2
Gene Accession No. P38935, P40694, Q9EQN5
Gene Alias AEP; antifreeze enhancer-binding protein; antifreeze enhancer-binding protein ortholog; ATP-dependent helicase IGHMBP2; cardiac transcription factor 1; CATF1; CMT2S; DNA-binding protein SMUBP-2; GF-1; Glial factor 1; HCSA; HMN6; Ighmbp2; immunoglobulin mu binding protein 2; immunoglobulin mu DNA binding protein 2; immunoglobulin mu-binding protein 2; immunoglobulin S mu binding protein 2; neuromuscular degeneration; nmd; p110 subunit; RIPE3b1; sma; SMARD1; Smbp2; Smbp-2; SMUBP2; ZFAND7; zinc finger, AN1-type domain 7
Gene Symbols IGHMBP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human IGHMBP2(Accession P38935), corresponding to amino acid residues E483-R533.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20589, 29532, 3508
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.