Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGIP Polyclonal Antibody
IGIP Polyclonal Antibody
Supplier: Thermo Scientific PA561783
Description
IGIP Polyclonal Antibody for IHC

Specifications
IGIP | |
Polyclonal | |
Unconjugated | |
IGIP | |
6330403M23Rik; C5orf53; IgA inducing protein; IgA-inducing protein; IgA-inducing protein homolog; IgA-inducing protein homolog (Bos taurus); IGIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
492311 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NJ69 | |
IGIP | |
Recombinant protein corresponding to Human IGIP. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction