Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGIP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983894
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3894 Supplier Bioss Supplier No. BS15574R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Enhances IgA secretion from B-cells stimulated via CD40.

Specifications

Antigen IGIP
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IGIP
Gene Accession No. A6NJ69
Gene Alias C5orf53; IgA inducing protein; IgA-inducing protein homolog; IgA-inducing protein homolog (Bos taurus); IGIP
Gene Symbols IGIP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGIP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 492311
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.