Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgJ Antibody (OTI3B3), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271016PEATT594
Description
IgJ Monoclonal specifically detects IgJ in Human samples. It is validated for Flow Cytometry.Specifications
IgJ | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
JCHAIN | |
Full length recombinant protein of human IgJ (NP_653247) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI3B3 | |
Flow Cytometry | |
IGCJJCH, immunoglobulin J chain, immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mupolypeptides | |
Mouse | |
Affinity Purified | |
Immunology | |
3512.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction