Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgJ Antibody (OTI3B3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271016PCP
Description
IgJ Monoclonal specifically detects IgJ in Human samples. It is validated for Flow Cytometry.Specifications
| IgJ | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| JCHAIN | |
| Full length recombinant protein of human IgJ (NP_653247) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI3B3 | |
| Flow Cytometry | |
| IGCJJCH, immunoglobulin J chain, immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mupolypeptides | |
| Mouse | |
| Affinity Purified | |
| Immunology | |
| 3512.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction