Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGKC Rabbit anti-Human, Unconjugated, Clone: KLC2886R, Abnova™

Catalog No. 89337876 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-876 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-876 Supplier Abnova Corporation Supplier No. RAB00451
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

IGKC Monoclonal antibody specifically detects IGKC in Human samples. It is validated for IHC-P, Flow Cyt.

Specifications

Antigen IGKC
Applications Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone KLC2886R
Conjugate Unconjugated
Description immunoglobulin kappa constant
Gene IGKC
Gene Alias HCAK1, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to full length human IGKC
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3514
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.