Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody
SDP

Rabbit Polyclonal Antibody

$182.00 - $401.00

Specifications

Antigen IgM
Applications Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018084
SDP
View Documents
Novus Biologicals
NB120171500.25ML
0.25 mL
Each for $182.00
Only null left
Add to Cart
 
NB12017150
SDP
View Documents
Novus Biologicals
NB12017150
1 mL
Each for $401.00
Only null left
Add to Cart
 
Description

Description

IgM Polyclonal specifically detects IgM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

IgM
Polyclonal
Purified
RUO
Human
3507
IgM isolated from human serum of a patient.
Primary
Store at 4°C.
Immunohistochemistry (Paraffin), Immunohistochemistry
Unconjugated
Rabbit
Immunology
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
IGHM
IgG
Affinity purified
This antibody reacts with the mu-chains of human IgM. Nonspecific antibodies have been removed by solid-phase adsorption.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.