Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Antibody (DA4-4 (same as SA-DA4 or HB57)), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234601PECY7
Description
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Flow Cytometry.Specifications
IgM | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
IGHM | |
Heavy chain of human IgM | |
0.1 mL | |
Primary | |
Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
DA4-4 (same as SA-DA4 or HB57) | |
Flow Cytometry | |
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
Mouse | |
Protein A or G purified | |
B Cell Development and Differentiation Markers, Immunology | |
3507.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction