Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Antibody (LO-MM-9), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP142328AF700
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Mouse IgM | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHM | |
The antibody reacts with the Mouse IgM antigen. | |
0.2 mL | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
LO-MM-9 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH | |
Rat | |
Protein A or G purified | |
RUO | |
Mouse mu heavy chain of immunoglobulin (determined by immunodot). Avidity on IgM: 7 * 10^8 M-1. No cross reactivity with other species. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction