Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody (LO-MM-9), Biotin, Novus Biologicals™
SDP

Catalog No. NBP142328B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142328B 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142328B Supplier Novus Biologicals Supplier No. NBP142328B
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Mouse IgM
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LO-MM-9
Conjugate Biotin
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Gene Symbols IGHM
Host Species Rat
Immunogen The antibody reacts with the Mouse IgM antigen.
Purification Method Protein A or G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Mouse mu heavy chain of immunoglobulin (determined by immunodot). Avidity on IgM: 7 * 10^8 M-1. No cross reactivity with other species.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.