Learn More
IgM Antibody (LO-MM-9), DyLight 350, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP142328UV
Catalog No. NBP142328UV
Only null left
Add to Cart
Description
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Mouse IgM
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
IGHM
The antibody reacts with the Mouse IgM antigen.
0.2 mL
Primary
Mouse
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
LO-MM-9
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Rat
Protein A or G purified
RUO
Mouse mu heavy chain of immunoglobulin (determined by immunodot). Avidity on IgM: 7 * 10^8 M-1. No cross reactivity with other species.
Store at 4C in the dark.
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.