Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody (SPM188) - IHC-Prediluted, Novus Biologicals™
SDP

Catalog No. NBP248240 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP248240 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP248240 Supplier Novus Biologicals Supplier No. NBP248240
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgM Monoclonal specifically detects IgM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IgM
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM188
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Gene Symbols IGHM
Host Species Mouse
Immunogen Heavy chain of human IgM
Purification Method Protein A or G purified
Quantity 7 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3507
Test Specificity Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Monomeric IgM is expressed as a membrane bound antibody on the surface of B cells and as a pentamer when secreted by plasma cells. IgM antibody is prominent in early immune responses to most antigens. Aberrant levels are associated with immune deficiency states, hereditary deficiencies, myeloma, Waldenstroms lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.