Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Antibody (SPM188), Novus Biologicals™

Mouse Monoclonal Antibody
$235.64 - $529.81
Specifications
| Antigen | IgM |
|---|---|
| Clone | SPM188 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24505500
![]() |
Novus Biologicals
NBP2450550.02MG |
0.02 mg |
Each for $235.64
|
|
|||||
NBP24505501
![]() |
Novus Biologicals
NBP2450550.1MG |
0.1 mg |
Each for $529.81
|
|
|||||
NBP24505502
![]() |
Novus Biologicals
NBP2450550.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| IgM | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 3507 | |
| Heavy chain of human IgM | |
| Primary | |
| Store at 4C. |
| SPM188 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| IGHM | |
| IgG1 κ | |
| Protein A or G purified | |
| Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Monomeric IgM is expressed as a membrane bound antibody on the surface of B cells and as a pentamer when secreted by plasma cells. IgM antibody is prominent in early immune responses to most antigens. Aberrant levels are associated with immune deficiency states, hereditary deficiencies, myeloma, Waldenstrom's macroglobulinemia, chronic infection and hepatocellular disease. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin s lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title