Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM Antibody (V53P3H2*B2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250439FR
Description
IgM Monoclonal specifically detects IgM in Fish samples. It is validated for ELISA.Specifications
IgM | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHM | |
Ovalbumin-conjugated synthetic peptide | |
0.1 mL | |
B Cell Development and Differentiation Markers, Immunology | |
3507 | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA | |
V53P3H2*B2 | |
ELISA | |
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Fish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction