Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM (Heavy Chain) Mouse anti-Human, Clone: SPM188, Abnova™
Mouse monoclonal antibody raised against native human IgM (Heavy Chain).
Supplier: Abnova Corporation MAB13096
Specifications
IgM (Heavy Chain) | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
DKFZp686I15196/DKFZp686I15212/FLJ00385/MGC104996/MGC52291/MU/VH | |
Mouse | |
Protein A/G purification | |
RUO | |
3507 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM188 | |
Mouse monoclonal antibody raised against native human IgM (Heavy Chain). | |
In 10mM PBS. | |
IGHM | |
Native purified human IgM (Heavy Chain). | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction