Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Kpa ItCl Rat, R718, Clone: R4-22, Isotype Control, BD Horizon™

Catalog No. BDB566950 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566950 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566950 Supplier BD Biosciences Supplier No. 566950
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The R4-22 monoclonal antibody has a Rat IgM, κ isotype and has no known specificity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM Kpa ItCl
Applications Flow Cytometry
Classification Monoclonal
Clone R4-22
Conjugate R718
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.